Assay Details
| Gene Symbol: CA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173898.2 | NP_776323.1 | 5 - 6 | 626 | 65 | |
| GenBank mRNA | X96503.1 | - | 626 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173898.2 | NP_776323.1 | 5 - 6 | 626 | 65 | |
| GenBank mRNA | X96503.1 | - | 626 | 65 | ||
Set Membership: |
|