Assay Details
| Gene Symbol: AGRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173983.2 | NP_776408.2 | 2 - 3 | 227 | 123 | |
| GenBank mRNA | AJ002025.1 | - | 2 - 3 | 227 | 123 | |
More Information
Set Membership: |
|

AGRP
Genomic Map