Assay Details
| Gene Symbol: AHSG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173984.3 | NP_776409.1 | 5 - 6 | 679 | 69 | |
| GenBank mRNA | BC147948.1 | - | 5 - 6 | 825 | 69 | |
| BC151430.1 | - | 5 - 6 | 742 | 69 | ||
| X16577.1 | - | 680 | 69 | |||
More Information
Set Membership: |
|

AHSG
Genomic Map