Assay Details
| Gene Symbol: LPAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174047.2 | NP_776472.1 | 2 - 3 | 813 | 74 | |
| GenBank mRNA | BC120114.1 | - | 3 - 4 | 1233 | 74 | |
| BC147965.1 | - | 3 - 4 | 1058 | 74 | ||
| U48236.1 | - | 2 - 3 | 813 | 74 | ||
More Information
Set Membership: |
|

LPAR1
Genomic Map