Assay Details
| Gene Symbol: FSHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174061.1 | NP_776486.1 | 9 - 10 | 923 | 98 | |
| GenBank mRNA | L22319.1 | - | 9 - 10 | 923 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FSHR| Gene Symbol: FSHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174061.1 | NP_776486.1 | 9 - 10 | 923 | 98 | |
| GenBank mRNA | L22319.1 | - | 9 - 10 | 923 | 98 | |
Set Membership: |
|