Assay Details
| Gene Symbol: EPYC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174307.2 | NP_776732.1 | 2 - 3 | 218 | 87 | |
| GenBank mRNA | U77127.1 | - | 2 - 3 | 218 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPYC| Gene Symbol: EPYC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174307.2 | NP_776732.1 | 2 - 3 | 218 | 87 | |
| GenBank mRNA | U77127.1 | - | 2 - 3 | 218 | 87 | |
Set Membership: |
|