Assay Details
| Gene Symbol: KCNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174371.2 | NP_776796.1 | 1 - 2 | 394 | 130 | |
| GenBank mRNA | AF286022.1 | - | 1 - 2 | 394 | 130 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174371.2 transcripts.
Set Membership: |
|

KCNA4
Genomic Map