Assay Details
| Gene Symbol: SFRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174460.2 | NP_776885.1 | 2 - 3 | 804 | 86 | |
| GenBank mRNA | U85945.1 | - | 2 - 3 | 804 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SFRP1| Gene Symbol: SFRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174460.2 | NP_776885.1 | 2 - 3 | 804 | 86 | |
| GenBank mRNA | U85945.1 | - | 2 - 3 | 804 | 86 | |
Set Membership: |
|