Assay Details
| Gene Symbol: EVC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174747.2 | NP_777172.1 | 10 - 11 | 1691 | 61 | |
| GenBank mRNA | AY028586.1 | - | 10 - 11 | 1691 | 61 | |
More Information
Set Membership: |
|

EVC
Genomic Map