Assay Details
| Gene Symbol: GCSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174844.1 | NP_777269.1 | 4 - 5 | 462 | 150 | |
| GenBank mRNA | BC120070.1 | - | 4 - 5 | 437 | 150 | |
| M58361.1 | - | 4 - 5 | 462 | 150 | ||
More Information
Set Membership: |
|

GCSH
Genomic Map