Assay Details
Gene Symbol: NDUFA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175794.2 | NP_786988.1 | 2 - 3 | 253 | 72 | |
GenBank mRNA | BC102993.1 | - | 2 - 3 | 262 | 72 | |
X63222.1 | - | 253 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
