Assay Details
| Gene Symbol: ARHGDIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175797.2 | NP_786991.1 | 5 - 6 | 447 | 92 | |
| GenBank mRNA | AF182001.1 | - | 5 - 6 | 447 | 92 | |
| BC102109.1 | - | 5 - 6 | 469 | 92 | ||
More Information
Set Membership: |
|

ARHGDIB
Genomic Map