Assay Details
| Gene Symbol: GLTP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175799.3 | NP_786993.1 | 2 - 3 | 172 | 83 | |
| GenBank mRNA | AF209701.1 | - | 2 - 3 | 173 | 83 | |
| BC105256.1 | - | 2 - 3 | 278 | 83 | ||
| BC142445.1 | - | 2 - 3 | 186 | 83 | ||
More Information
Set Membership: |
|

GLTP
Genomic Map