Assay Details
| Gene Symbol: NDUFA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175815.2 | NP_787009.1 | 1 - 2 | 133 | 89 | |
| GenBank mRNA | BC118428.1 | - | 1 - 2 | 120 | 89 | |
| X63219.1 | - | 133 | 89 | |||
More Information
Set Membership: |
|

NDUFA2
Genomic Map