Assay Details
| Gene Symbol: COX7A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176674.2 | NP_788847.1 | 1 - 2 | 77 | 72 | |
| GenBank mRNA | BC114907.1 | - | 1 - 2 | 55 | 72 | |
| X56739.1 | - | 77 | 72 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC114907.1, NM_176674.2, X56739.1 transcripts.
Set Membership: |
|

COX7A1
Genomic Map