Assay Details
| Gene Symbol: COX7A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176674.2 | NP_788847.1 | 2 - 3 | 157 | 74 | |
| GenBank mRNA | BC114907.1 | - | 2 - 3 | 135 | 74 | |
| X56739.1 | - | 157 | 74 | |||
More Information
Set Membership: |
|

COX7A1
Genomic Map