Assay Details
| Gene Symbol: FLT3LG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181030.2 | NP_851373.1 | 2 - 3 | 150 | 64 | |
| GenBank mRNA | AB051841.1 | - | 2 - 3 | 150 | 64 | |
| AF282985.1 | - | 2 - 3 | 150 | 64 | ||
| AF282986.1 | - | 2 - 3 | 150 | 64 | ||
| BC133533.1 | - | 3 - 4 | 226 | 64 | ||
More Information
Set Membership: |
|

FLT3LG
Genomic Map