Assay Details
Gene Symbol: FLT3LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181030.2 | NP_851373.1 | 3 - 4 | 201 | 64 | |
GenBank mRNA | AB051841.1 | - | 3 - 4 | 201 | 64 | |
AF282985.1 | - | 3 - 4 | 201 | 64 | ||
AF282986.1 | - | 3 - 4 | 201 | 64 | ||
BC133533.1 | - | 4 - 5 | 277 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
