Assay Details
| Gene Symbol: B3GAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014393.1 | NP_001014415.1 | 3 - 4 | 916 | 134 | |
| GenBank mRNA | AJ890839.1 | - | 3 - 4 | 916 | 134 | |
More Information
Set Membership: |
|

B3GAT1
Genomic Map