Assay Details
| Gene Symbol: TCAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014915.2 | NP_001014915.1 | 1 - 2 | 115 | 63 | |
| GenBank mRNA | BC103048.1 | - | 1 - 2 | 136 | 63 | |
| BC142094.1 | - | 1 - 2 | 136 | 63 | ||
More Information
Set Membership: |
|

TCAP
Genomic Map