Assay Details
| Gene Symbol: METTL21E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014922.1 | NP_001014922.1 | 1 - 2 | 282 | 110 | |
| GenBank mRNA | BC149818.1 | - | 1 - 2 | 122 | 110 | |
| BT021670.1 | - | 1 - 2 | 282 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149818.1, NM_001014922.1 transcripts.
Set Membership: |
|

METTL21E
Genomic Map