Assay Details
| Gene Symbol: METTL21E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014922.1 | NP_001014922.1 | 4 - 5 | 657 | 119 | |
| GenBank mRNA | BC149818.1 | - | 4 - 5 | 497 | 119 | |
| BT021670.1 | - | 4 - 5 | 657 | 119 | ||
More Information
Set Membership: |
|

METTL21E
Genomic Map