Assay Details
| Gene Symbol: OTUB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015517.1 | NP_001015517.1 | 4 - 5 | 337 | 63 | |
| GenBank mRNA | BT020786.1 | - | 4 - 5 | 337 | 63 | |
More Information
Set Membership: |
|

OTUB2
Genomic Map