Assay Details
| Gene Symbol: SLC25A21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015587.1 | NP_001015587.1 | 5 - 6 | 574 | 62 | |
| GenBank mRNA | BC126566.1 | - | 5 - 6 | 714 | 62 | |
| BT020902.1 | - | 5 - 6 | 574 | 62 | ||
More Information
Set Membership: |
|

SLC25A21
Genomic Map