Assay Details
| Gene Symbol: EMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024562.1 | NP_001019733.1 | 2 - 3 | 433 | 71 | |
| GenBank mRNA | BC126787.1 | - | 2 - 3 | 217 | 71 | |
| BT021531.1 | - | 2 - 3 | 433 | 71 | ||
More Information
Set Membership: |
|

EMP3
Genomic Map