Assay Details
| Gene Symbol: NLRP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024664.1 | NP_001019835.1 | 7 - 8 | 2686 | 114 | |
| GenBank mRNA | DQ011673.1 | - | 7 - 8 | 2686 | 114 | |
| DQ092866.1 | - | 7 - 8 | 2686 | 114 | ||
More Information
Set Membership: |
|

NLRP9
Genomic Map