Assay Details
Gene Symbol: TICAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030301.1 | NP_001025472.1 | 2 | 1249 | 115 | |
GenBank mRNA | AJ879588.1 | - | 2 | 1249 | 115 | |
BC151622.1 | - | 3 | 1521 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
