Assay Details
| Gene Symbol: UBTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034261.2 | NP_001029433.1 | 2 - 3 | 442 | 98 | |
| GenBank mRNA | BC103175.1 | - | 2 - 3 | 443 | 98 | |
More Information
Set Membership: |
|

UBTD1
Genomic Map