Assay Details
| Gene Symbol: CNEP1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034303.2 | NP_001029475.1 | 3 - 4 | 303 | 72 | |
| GenBank mRNA | BC103188.1 | - | 3 - 4 | 304 | 72 | |
More Information
Set Membership: |
|

CNEP1R1
Genomic Map