Assay Details
| Gene Symbol: TMEM106B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034337.1 | NP_001029509.1 | 2 - 3 | 419 | 64 | |
| GenBank mRNA | BC102965.1 | - | 2 - 3 | 419 | 64 | |
More Information
Set Membership: |
|

TMEM106B
Genomic Map