Assay Details
| Gene Symbol: MBD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034405.2 | NP_001029577.1 | 6 - 7 | 1596 | 74 | |
| GenBank mRNA | BC103441.1 | - | 6 - 7 | 1597 | 74 | |
More Information
Set Membership: |
|

MBD4
Genomic Map