Assay Details
| Gene Symbol: ZSCAN16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034411.1 | NP_001029583.1 | 3 - 4 | 659 | 77 | |
| GenBank mRNA | BC103279.1 | - | 3 - 4 | 659 | 77 | |
More Information
Set Membership: |
|

ZSCAN16
Genomic Map