Assay Details
Gene Symbol: TIMM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034498.1 | NP_001029670.1 | 1 - 2 | 266 | 88 | |
GenBank mRNA | BC148116.1 | - | 1 - 2 | 268 | 88 | |
BT021190.1 | - | 1 - 2 | 266 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
