Assay Details
| Gene Symbol: TMUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034675.1 | NP_001029847.1 | 1 - 2 | 37 | 92 | |
| GenBank mRNA | BC103270.1 | - | 1 - 2 | 37 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103270.1, NM_001034675.1 transcripts.
Set Membership: |
|

TMUB1
Genomic Map