Assay Details
| Gene Symbol: OSBPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035020.2 | NP_001030192.1 | 2 - 3 | 209 | 61 | |
| GenBank mRNA | BC102883.1 | - | 2 - 3 | 210 | 61 | |
| BT021611.1 | - | 2 - 3 | 211 | 61 | ||
More Information
Set Membership: |
|

OSBPL2
Genomic Map