Assay Details
| Gene Symbol: MYG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035038.1 | NP_001030210.1 | 2 - 3 | 329 | 80 | |
| GenBank mRNA | BC111153.1 | - | 2 - 3 | 348 | 80 | |
| BT021616.1 | - | 2 - 3 | 342 | 80 | ||
| BT021636.1 | - | 2 - 3 | 336 | 80 | ||
| BT021764.1 | - | 2 - 3 | 329 | 80 | ||
More Information
Set Membership: |
|

MYG1
Genomic Map