Assay Details
| Gene Symbol: MYG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035038.1 | NP_001030210.1 | 3 - 4 | 491 | 67 | |
| GenBank mRNA | BC111153.1 | - | 3 - 4 | 510 | 67 | |
| BT021636.1 | - | 3 - 4 | 498 | 67 | ||
| BT021764.1 | - | 3 - 4 | 491 | 67 | ||
More Information
Set Membership: |
|

MYG1
Genomic Map