Assay Details
| Gene Symbol: MYG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035038.1 | NP_001030210.1 | 6 - 7 | 947 | 59 | |
| GenBank mRNA | BC111153.1 | - | 6 - 7 | 966 | 59 | |
| BT021616.1 | - | 6 - 7 | 960 | 59 | ||
| BT021636.1 | - | 6 - 7 | 954 | 59 | ||
| BT021764.1 | - | 6 - 7 | 947 | 59 | ||
More Information
Set Membership: |
|

MYG1
Genomic Map