Assay Details
| Gene Symbol: CYP4F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035042.1 | NP_001030214.1 | 2 - 3 | 266 | 82 | |
| GenBank mRNA | BC102056.1 | - | 2 - 3 | 266 | 82 | |
More Information
Set Membership: |
|

CYP4F2
Genomic Map