Assay Details
| Gene Symbol: TMEM14C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035437.1 | NP_001030514.1 | 4 - 5 | 277 | 62 | |
| GenBank mRNA | BC102229.1 | - | 4 - 5 | 277 | 62 | |
More Information
Set Membership: |
|

TMEM14C
Genomic Map