Assay Details
| Gene Symbol: SPATA19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076498.2 | NP_001069966.1 | 4 - 5 | 386 | 68 | |
| GenBank mRNA | BC102751.1 | - | 4 - 5 | 387 | 68 | |
More Information
Set Membership: |
|

SPATA19
Genomic Map