Assay Details
| Gene Symbol: TLR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174197.2 | NP_776622.1 | 1 - 2 | 177 | 109 | |
| GenBank mRNA | AF368419.1 | - | 1 - 2 | 177 | 109 | |
| AY634629.1 | - | 1 - 2 | 117 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174197.2 transcripts.
Set Membership: |
|

TLR2
Genomic Map