Assay Details
| Gene Symbol: CKMT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174275.2 | NP_776700.1 | 5 - 6 | 921 | 68 | |
| GenBank mRNA | AB003307.1 | - | 5 - 6 | 921 | 68 | |
| BC102422.1 | - | 6 - 7 | 68 | |||
More Information
Set Membership: |
|

CKMT1A
Genomic Map