Assay Details
| Gene Symbol: HSD17B10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174334.3 | NP_776759.1 | 2 - 3 | 210 | 57 | |
| GenBank mRNA | AB002156.1 | - | 2 - 3 | 211 | 57 | |
| BC110264.1 | - | 2 - 3 | 208 | 57 | ||
More Information
Set Membership: |
|

HSD17B10
Genomic Map