Assay Details
| Gene Symbol: HSD17B10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174334.3 | NP_776759.1 | 3 - 4 | 372 | 73 | |
| GenBank mRNA | AB002156.1 | - | 3 - 4 | 373 | 73 | |
| BC110264.1 | - | 3 - 4 | 370 | 73 | ||
More Information
Set Membership: |
|

HSD17B10
Genomic Map