Assay Details
| Gene Symbol: FCGR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174538.2 | NP_776963.1 | 3 - 4 | 320 | 66 | |
| GenBank mRNA | AF162866.1 | - | 3 - 4 | 320 | 66 | |
| BC112691.1 | - | 3 - 4 | 332 | 66 | ||
More Information
Set Membership: |
|

FCGR1A
Genomic Map