Assay Details
| Gene Symbol: FUT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174735.2 | NP_777160.1 | 1 - 2 | 66 | 72 | |
| GenBank mRNA | AF531093.1 | - | 1 - 2 | 66 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174735.2 transcripts.
Set Membership: |
|

FUT9
Genomic Map