Assay Details
| Gene Symbol: COX7A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175807.1 | NP_787001.1 | 1 - 2 | 88 | 93 | |
| GenBank mRNA | BC102664.1 | - | 1 - 2 | 109 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102664.1, NM_175807.1 transcripts.
Set Membership: |
|

COX7A2
Genomic Map