Assay Details
| Gene Symbol: COX7A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175807.1 | NP_787001.1 | 3 - 4 | 275 | 82 | |
| GenBank mRNA | BC102664.1 | - | 3 - 4 | 296 | 82 | |
| MK423994.1 | - | 3 - 4 | 207 | 82 | ||
More Information
Set Membership: |
|

COX7A2
Genomic Map