Assay Details
Gene Symbol: JPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034734.2 | NP_001029906.1 | 4 - 5 | 431 | 111 | |
GenBank mRNA | BC102268.1 | - | 4 - 5 | 434 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
